missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIHBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
200.00 € - 450.00 €
Specifications
| Antigen | GPIHBP1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18250751
|
Novus Biologicals
NB110-55293SS |
0.025 mL |
200.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18790214
|
Novus Biologicals
NB110-55293 |
0.1 mL |
450.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPIHBP1 Polyclonal specifically detects GPIHBP1 in Human samples. It is validated for Western Blot.Specifications
| GPIHBP1 | |
| Western Blot 2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
| GPIHBP1 | |
| IgG | |
| Affinity Purified | |
| Human. Immunogen has 78% homology to mouse. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8IV16 | |
| 338328 | |
| A synthetic peptide corresponding to an internal region [within residues 100-180] of human GPIHBP1. [Swiss-Prot# Q8IV16] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title