missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR17 Antibody (CL10474) [Alexa Fluor« 405], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
GPR17 Monoclonal antibody specifically detects GPR17 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | GPR17 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL10474 |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686M18273, G protein-coupled receptor 17, G-protein coupled receptor 17, P2Y-like receptor, R12, UDP/CysLT receptor, uracil nucleotide/cysteinyl leukotriene receptor |
| Host Species | Mouse |
| Immunogen | This antibody was generated using a synthetic peptide of Q13304, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?