missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GPR4 Antibody, Novus Biologicals™

Product Code. p-200085333 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30066915 100 μg 100µg
30067128 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30066915 Supplier Novus Biologicals™ Supplier No. NBP330859100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR4 Polyclonal antibody specifically detects GPR4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 1:400
Formulation Phosphate buffered saline
Gene Alias G protein-coupled receptor 4, GPR19, G-protein coupled receptor 19, G-protein coupled receptor 4
Host Species Rabbit
Immunogen Synthetic peptide derived from the GPR4 protein.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2828
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.