missing translation for 'onlineSavingsMsg'
Learn More

GPR56 Antibody (2446B), mFluor Violet 450 SE, R&D Systems™

Product Code. 18620310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620310 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18620310 Supplier Novus Biologicals Supplier No. FAB46361MFV450
  • ONLINE EXCLUSIVE PRICE
    668.09 €
    574.35 € / 0.1mL
    Save 93.74 €
    14% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

GPR56 Monoclonal specifically detects GPR56 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR56
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2446B
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BFPP, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4G-protein coupled receptor 56, TM7XN1DKFZp781L1398
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val345, Accession # Q9Y653
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9289
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.