missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Granzyme B Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Granzyme B Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1157-S1

 View more versions of this product

Product Code. 12659136

  • ONLINE EXCLUSIVE PRICE
    499.00 €
    419.04 € / 500µL
    Save 79.96 €
    16% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified. One of these, human granzyme B, is involved in target cell apoptosis during lymphocyte-mediated cytotoxicity. Exocytosis of granzyme-containing granules in the cytoplasm of the target cell leads to the induction of DNA fragmentation and apoptosis.

Host Species: Mouse

Clone: GZB01

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein encoding the N-terminus of the mature granzyme B

Molecular Weight: 32 kDa

Positive Control: Hodgkin's lymphoma, Tonsil

Cellular Localization: Granular cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Granzyme B Ab-1
Monoclonal
Unconjugated
Recombinant protein encoding the N-terminus of the mature granzyme B
500 μL
Apoptosis
Human
IgG2a
Immunohistochemistry (Paraffin)
GZB01
Mouse
Hodgkin's lymphoma
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.