missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Granzyme B Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Granzyme B Ab-1, Mouse Monoclonal Antibody.

142.80 € - 627.53 €

Specifications

Antigen Granzyme B Ab-1
Clone GZB01
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12649136
View Documents
Epredia™
MS-1157-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12659136
View Documents
Epredia™
MS-1157-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
499.00 €
419.04 € / 500µL
Save 79.96 €
16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12639136
View Documents
Epredia™
MS-1157-S
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
755.00 €
627.53 € / 1mL
Save 127.47 €
16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12629136
View Documents
Epredia™
MS-1157-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12619136
View Documents
Epredia™
MS-1157-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified. One of these, human granzyme B, is involved in target cell apoptosis during lymphocyte-mediated cytotoxicity. Exocytosis of granzyme-containing granules in the cytoplasm of the target cell leads to the induction of DNA fragmentation and apoptosis.

Host Species: Mouse

Clone: GZB01

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein encoding the N-terminus of the mature granzyme B

Molecular Weight: 32 kDa

Positive Control: Hodgkin's lymphoma, Tonsil

Cellular Localization: Granular cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Specifications

Specifications

Granzyme B Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Recombinant protein encoding the N-terminus of the mature granzyme B
GZB01
Monoclonal
Mouse
Apoptosis
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.