missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB7 Antibody (SR2057), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21932-100ul
This item is not returnable.
View return policy
Description
GRB7 Monoclonal antibody specifically detects GRB7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| GRB7 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2057 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500 | |
| B47, Epidermal growth factor receptor GRB-7, GRB7 adapter protein, growth factor receptor-bound protein 7 | |
| A synthesized peptide derived from human GRB7 (Uniprot #: Q14451) | |
| 100 μg | |
| Breast Cancer, Cancer, Extracellular Matrix | |
| 2886 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction