missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRB7 Antibody (SR2057), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | GRB7 |
|---|---|
| Clone | SR2057 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659194
|
Novus Biologicals
NBP3-21932-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672114
|
Novus Biologicals
NBP3-21932-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRB7 Monoclonal antibody specifically detects GRB7 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| GRB7 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| B47, Epidermal growth factor receptor GRB-7, GRB7 adapter protein, growth factor receptor-bound protein 7 | |
| A synthesized peptide derived from human GRB7 (Uniprot #: Q14451) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2057 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Extracellular Matrix | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2886 | |
| IgG | |
| Affinity purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit