missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GroEL Antibody (9A1/2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-97895-0.05MG
This item is not returnable.
View return policy
Description
GroEL Monoclonal specifically detects GroEL in E. coli samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| GroEL | |
| Monoclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 948665 | |
| E. coli | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 9A1/2 | |
| Western Blot 1:10000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
| Cpn60 chaperonin GroEL, large subunit of GroESL, ECK4137, groEL, JW4103, mopA | |
| E. coli GroEL. | |
| 0.05 mg | |
| Primary | |
| Recognizes Bacteria GroEL. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction