missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GroEL Antibody (9A1/2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
193.00 € - 650.00 €
Specifications
| Antigen | GroEL |
|---|---|
| Clone | 9A1/2 |
| Dilution | Western Blot 1:10000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 |
| Applications | Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075399
|
Novus Biologicals
NBP1-97895-0.025MG |
0.025 mg |
193.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18044649
|
Novus Biologicals
NBP1-97895-0.05MG |
0.05 mg |
418.00 €
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18098699
|
Novus Biologicals
NBP1-97895-0.2MG |
0.2 mg |
650.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GroEL Monoclonal specifically detects GroEL in E. coli samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| GroEL | |
| Western Blot 1:10000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| 948665 | |
| IgG1 | |
| Protein G purified | |
| Recognizes Bacteria GroEL. |
| 9A1/2 | |
| Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| E. coli | |
| Cpn60 chaperonin GroEL, large subunit of GroESL, ECK4137, groEL, JW4103, mopA | |
| E. coli GroEL. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title