missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81943
This item is not returnable.
View return policy
Description
GSK-3 alpha Polyclonal specifically detects GSK-3 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| GSK-3 alpha | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
| Rabbit | |
| 51 kDa | |
| 0.1 mL | |
| Cell Biology, Hypoxia, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 2931 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100∼1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunofluorescence 1:100-1:200 | |
| P49840 | |
| GSK3A | |
| The antiserum was produced against synthesized non-phosphopeptide derived from human GSK-3 alpha around amino acids 19∼23 (T-S-S-F-A). IP data from customer review. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of total GSK-3 alpha protein. Does not detect GSK-3 beta protein | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction