missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00 € - 491.00 €
Specifications
| Antigen | GSK-3 alpha |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18226995
|
Novus Biologicals
NB100-81943-0.025ML |
0.025 mL |
189.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18238270
|
Novus Biologicals
NB100-81943 |
0.1 mL |
491.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSK-3 alpha Polyclonal specifically detects GSK-3 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| GSK-3 alpha | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
| GSK3A | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cell Biology, Hypoxia, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| P49840 | |
| 2931 | |
| IgG | |
| Affinity Purified | |
| 51 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title