missing translation for 'onlineSavingsMsg'
Learn More

GTF2IRD1 Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30508990 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30508990 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30508990 Supplier Novus Biologicals Supplier No. NBP338300AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GTF2IRD1 Polyclonal antibody specifically detects GTF2IRD1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2IRD1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias BEN, CREAM1, General transcription factor III, general transcription factor II-I repeat domain-containing protein 1, GTF2I repeat domain containing 1, GTF2I repeat domain-containing protein 1, GTF3GTF2I repeat domain-containing 1, hMusTRD1alpha1, Muscle TFII-I repeat domain-containing protein 1, muscle TFII-I repeat domain-containing protein 1 alpha 1, MusTRD1, MusTRD1/BEN, MUSTRD1general transcription factor 3, RBAP2WBSCR12binding factor for early enhancer, Slow-muscle-fiber enhancer-binding protein, USE B1-binding protein, WBS, WBSCR11, Williams-Beuren syndrome chromosomal region 11 protein, Williams-Beuren syndrome chromosomal region 12 protein, Williams-Beuren syndrome chromosome region 11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 660-959 of human GTF2IRD1 (NP_057412.1).,, Sequence:, SLGFSPPALPPERDSGDPLVDESLKRQGFQENYDARLSRIDIANTLREQVQDLFNKKYGEALGIKYPVQVPYKRIKSNPGSVIIEGLPPGIPFRKPCTFGSQNLERILAVADKIKFTVTRPFQGLIPKPDEDDANRLGEKVILREQVKELFNEKYGEALGLNRPVLVPYKLIRDSPDAVEVTGLPDDIPFRNPNTYDIHRLEKILKAREHVRMVIINQLQPFAEICNDAKVPAKDSSIPKRKRKRVSEGNSVSSSSSSSSSSSSNPDSVASANQISLVQWPMYMVDYAGLNVQLPGPLNY
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9569
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.