missing translation for 'onlineSavingsMsg'
Learn More

GTP binding protein era homolog Antibody [HRP], Novus Biologicals Biologicals™

Product Code. 30501880 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30501880 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30501880 Supplier Novus Biologicals Supplier No. NBP335133H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GTP binding protein era homolog Polyclonal antibody specifically detects GTP binding protein era homolog in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTP binding protein era homolog
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CEGA, Conserved ERA-like GTPase, ERA, Era (E. coli G-protein homolog)-like 1, Era G-protein-like 1 (E. coli), ERAL1A, ERA-like protein 1, ERA-W, GTPase Era, mitochondrial, GTPase, human homolog of E. coli essential cell cycle protein Era, GTP-binding protein era homolog, HERA, H-ERA, HERA-A, HERA-B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 198-437 of human GTP binding protein era homolog (NP_005693.1).,, Sequence:, DLVVVLVDVSDKWTRNQLSPQLLRCLTKYSQIPSVLVMNKVDCLKQKSVLLELTAALTEGVVNGKKLKMRQAFHSHPGTHCPSPAVKDPNTQSVGNPQRIGWPHFKEIFMLSALSQEDVKTLKQYLLTQAQPGPWEYHSAVLTSQTPEEICANIIREKLLEHLPQEVPYNVQQKTAVWEEGPGGELVIQQKLLVPKESYVKLLIGPKGHVISQIAQEAGHDLMDIFLCDVDIRLSVKLLK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26284
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.