missing translation for 'onlineSavingsMsg'
Learn More

GTPBP5 Antibody [Alexa Fluor™ 488], Novus Biologicals™

Product Code. 30566144 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30566144 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30566144 Supplier Novus Biologicals Supplier No. NBP346156AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GTPBP5 Polyclonal antibody specifically detects GTPBP5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifica

Antigen GTPBP5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias dJ1005F21.2, FLJ10741, GTP binding protein 5 (putative), GTP-binding protein 5, GTP-binding protein 5 (putative), MGC29512, ObgH1OBGH1, Protein obg homolog 1
Host Species Rabbit
Immunogen GTP binding protein 5(putative). (Uniprot# Q9H4K7)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26164
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.