missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Halcinonide

Product Code. 16637022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16637022 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16637022 Supplier Thermo Scientific Chemicals Supplier No. 464980010
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Packaging glass bottle
CAS 3093-35-4
Certificate of Analysis Available
Shipping Condition normal conditions
Physical Form Powder
Molecular Formula C24H32ClFO5
MDL Number MFCD00866006
Color White to Yellow
Melting Point 264°C to 265°C
Solubility Soluble in acetone, chloroform and DMSO; Slightly soluble in benzene, ethanol, methanol and ethyl ether; Insoluble in hexanes and water
InChI Key MUQNGPZZQDCDFT-JNQJZLCISA-N
Type Reagent
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CCl
IUPAC Name (1S,2S,4R,8S,9S,11S,12R,13S)-8-(2-chloroacetyl)-12-fluoro-11-hydroxy-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icos-17-en-16-one
Molecular Weight (g/mol) 454.96
Quantity 1 g
Formula Weight 454.96
Chemical Name or Material Halcinonide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.