missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol, LoGiCal

Product Code. 30464132
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464132 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464132 Supplier LoGiCal Supplier No. LGCFOR1165.00
 Restricted Item
Estimated Shipment: 30-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Haloperidol
CAS 52-86-8
Analyte or Component Names Haloperidol
Shipping Condition Room Temperature
Molecular Formula C21 H23 Cl F N O2
Certifications/Compliance ISO 9001
Synonym 1-Butanone, 4-[4-(4-chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-, Butyrophenone, 4-[4-(p-chlorophenyl)-4-hydroxypiperidino]-4'-fluoro- (6CI,8CI), 1-(3-p-Fluorobenzoylpropyl)-4-p-chlorophenyl-4-hydroxypiperidine, 4-(4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl)-1-(4-fluorophenyl)-1-butanone, 4-(4-Hydroxy-4'-chloro-4-phenylpiperidino)-4'-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4'-fluorobutyrophenone, Aloperidin, Bioperidolo, Brotopon, Dozic, Einalon S, Eukystol, Haldol, Haloperidol, Haloperin, Halosten, Keselan, Linton, McN-JR 1625, NSC 170973, NSC 615296, Neurodol, Peluces, R 1625, Serenace, Serenase, Serenelfi, Sigaperidol, g-[4-(p-Chlorphenyl)-4-hydroxypiperidino]-p-fluorbutyrophenone, g-[4-(p-Chlorphenyl)-4-hydroxypiperidino]-p-fluorbutyrophenone, Haloperidol Decanoate Impurity G
InChI Formula InChI=1S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2
Solution Type Neat
SMILES OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 375.86
Quantity 10 mg
Formula Weight 375.1401 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.