Learn More
Hamilton™ Kel-F Hub Side Port Needles (Luer Lock)
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51mm). Custom lengths and point styles are available.
184.00 € - 188.00 €
Specifications
Material | Nickel-plated Brass (Hub) |
---|---|
Length (English) Needle | 2 in. |
Product Type | Kel-F Hub Needle |
Needle Point Style | Cone |
For Use With (Equipment) | Luer-tip and PTFE Luer-lock Syringes and Connectors |
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
10318662
|
Hamilton™
7746-01 |
16 G |
184.00 €
Pack of 6 |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10135452
|
Hamilton™
7746-07 |
23 G |
184.00 €
Pack of 6 |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11540152
|
Hamilton™
7746-03 |
18 G |
184.00 €
Pack of 6 |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11550152
|
Hamilton™
7746-04 |
20 G |
184.00 €
Pack of 6 |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10245612
|
Hamilton™
7746-06 |
22 G |
184.00 €
Pack of 6 |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11570152
|
Hamilton™
7746-08 |
24 G |
184.00 €
Pack of 6 |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11590152
|
Hamilton™
7746-10 |
26 G |
184.00 €
Pack of 6 |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11500162
|
Hamilton™
7746-11 |
22s G |
184.00 €
Pack of 6 |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10217734
|
Hamilton™
7746-12 |
26s G |
184.00 €
Pack of 6 |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11580152
|
Hamilton™
7746-09 |
25 G |
188.00 €
Pack of 6 |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11560152
|
Hamilton™
7746-05 |
21 G |
188.00 €
Pack of 6 |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11530152
|
Hamilton™
7746-02 |
17 G |
184.00 €
Pack of 6 |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Kel-F Hub Compatible Syringe Terminations
- Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
- PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
- Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
Six needles per pack.
Specifications
Nickel-plated Brass (Hub) | |
Kel-F Hub Needle | |
Luer-tip and PTFE Luer-lock Syringes and Connectors |
2 in. | |
Cone |