missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB-EGF Antibody (MM0324-3R44) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12352-0.025MG
This item is not returnable.
View return policy
Description
HB-EGF Monoclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.
Specifications
| HB-EGF | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1000, Immunohistochemistry-Paraffin 1:50-1:200, Blocking/Neutralizing 50-200 molar excess | |
| Q53H93 | |
| HBEGF | |
| Human HB-EGF | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Blocking Assay, Immunohistochemistry (Paraffin) | |
| MM0324-3R44 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1839 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction