missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HB-EGF Antibody (MM0324-3R44) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
213.00 € - 507.00 €
Specifications
| Antigen | HB-EGF |
|---|---|
| Clone | MM0324-3R44 |
| Concentration | LYOPH |
| Applications | Western Blot, Blocking Assay, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18082816
|
Novus Biologicals
NBP2-12352-0.025MG |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18771374
|
Novus Biologicals
NBP2-12352 |
0.1 mg | |||||||
Description
HB-EGF Monoclonal specifically detects HB-EGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
| HB-EGF | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q53H93 | |
| 1839 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0324-3R44 | |
| Western Blot, Blocking Assay, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
| HBEGF | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title