missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-56343SS
This item is not returnable.
View return policy
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| HDAC6 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| In NIH 3T3, a 134 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 2 - 5 ug/ml, Simple Western 1:20, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Imbriano (2005)), Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation assay dependent, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 | |
| NP_002495 | |
| HDAC6 | |
| Synthetic peptide made to an N-terminal portion of human HDAC6 (between amino acids 1-50) [UniProt Q9UBN7] | |
| 0.025 mg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research | |
| 10013 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction