missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
231.00 € - 548.00 €
Specifications
| Antigen | HDAC6 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 - 5 ug/ml, Simple Western 1:20, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Imbriano (2005)), Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation assay dependent, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 |
| Applications | Western Blot, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18090926
|
Novus Biologicals
NB100-56343SS |
0.025 mg |
231.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745563
|
Novus Biologicals
NB100-56343 |
0.1 mg |
548.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| HDAC6 | |
| Western Blot 2 - 5 ug/ml, Simple Western 1:20, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Imbriano (2005)), Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation assay dependent, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| NP_002495 | |
| 10013 | |
| Synthetic peptide made to an N-terminal portion of human HDAC6 (between amino acids 1-50) [UniProt Q9UBN7] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research | |
| PBS with 0.02% Sodium Azide | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| HDAC6 | |
| IgG | |
| Immunogen affinity purified | |
| In NIH 3T3, a 134 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title