missing translation for 'onlineSavingsMsg'
Learn More

HDJ2 Antibody (SR1766), Novus Biologicals™

Product Code. 18686894 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18686894 50 μg 50µL
18609623 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18686894 Supplier Novus Biologicals Supplier No. NBP32227050ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HDJ2 Monoclonal antibody specifically detects HDJ2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDJ2
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone SR1766
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias dj-2, DjA1, DnaJ (Hsp40) homolog, subfamily A, member 1, dnaJ homolog subfamily A member 1, DnaJ protein homolog 2, DNAJ2, hDJ-2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, heat shock protein, DNAJ-like 2, HSDJ, HSJ2, HSJ-2, HSPF4hdj-2, Human DnaJ protein 2, NEDD7, neural precursor cell expressed, developmentally down-regulated 7
Host Species Rabbit
Immunogen A synthesized peptide derived from human HDJ2 (Uniprot #: P31689)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3301
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.