Learn More
HELIOS Mouse, anti-Human, Clone: 1F1F10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68558-1-IG-150UL
This item is not returnable.
View return policy
Description
The HELIOS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HELIOS. This antibody recognizes human antigen. The HELIOS antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| HELIOS | |
| 1F1F10 | |
| HELIOS, IKZF2, Zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
| 150 μL | |
| 22807 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunoprecipitation, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2b |