missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HELIOS Mouse, anti-Human, Clone: 1F1F10, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | HELIOS |
|---|---|
| Clone | 1F1F10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18080516
|
Proteintech
68558-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040736
|
Proteintech
68558-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HELIOS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HELIOS. This antibody recognizes human antigen. The HELIOS antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| HELIOS | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| HELIOS, IKZF2, Zinc finger protein Helios, ZNF1A2, ZNFN1A2 | |
| Primary |
| 1F1F10 | |
| Western Blot, Immunoprecipitation, ELISA | |
| Mouse | |
| 22807 | |
| IgG2b | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title