missing translation for 'onlineSavingsMsg'
Learn More

Hemoglobin epsilon Antibody (6G7) [PE], Novus Biologicals Biologicals™

Product Code. 30520419 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520419 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520419 Supplier Novus Biologicals Supplier No. NBP342981PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hemoglobin epsilon Monoclonal antibody specifically detects Hemoglobin epsilon in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemoglobin epsilon
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6G7
Conjugate PE
Formulation PBS
Gene Alias epsilon globin, Epsilon-globin, HBEepsilon-globin, Hemoglobin epsilon chain, hemoglobin subunit epsilon, hemoglobin, epsilon 1
Host Species Mouse
Immunogen hemoglobin, epsilon 1 - (Uniprot# P02100)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3046
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.