missing translation for 'onlineSavingsMsg'
Learn More

Hemoglobin mu Antibody [FITC], Novus Biologicals™

Product Code. 30566161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30566161 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30566161 Supplier Novus Biologicals Supplier No. NBP346145F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hemoglobin mu Polyclonal antibody specifically detects Hemoglobin mu in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemoglobin mu
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias alpha globin pseudogene 2, HBAP2, Hemoglobin mu chain, hemoglobin subunit mu, hemoglobin, alpha pseudogene 2, hemoglobin, mu, mu-globin
Host Species Rabbit
Immunogen hemoglobin, mu. (Uniprot# Q6B0K9)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3042
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.