missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu Ab-17 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (For cytoplasmic), Epredia™ HER-2/c-erbB-2/neu Ab-17, Mouse Monoclonal Antibody provides accurate, reproducible results..

Brand:  Epredia MS-730-R7

 View more versions of this product

Product Code. 12682187

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: e2-4001 + 3B5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Cytoplasmic

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human HER-2 protein (3B5)

Molecular Weight: 185kDa

Positive Control: LS174T cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (For cytoplasmic)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HER-2/c-erbB-2/neu Ab-17
Monoclonal
Mouse
7 mL
EGFR Family
Human, Rat
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
e2-4001 + 3B5
Unconjugated
Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5)
ASR, IVD
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.