missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu Ab-17 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (For cytoplasmic), Epredia™ HER-2/c-erbB-2/neu Ab-17, Mouse Monoclonal Antibody provides accurate, reproducible results..

161.00 € - 767.00 €

Specifications

Antigen HER-2/c-erbB-2/neu Ab-17
Clone e2-4001 + 3B5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Form Concentration Price Quantity & Availability  
12622187
View Documents
Epredia
MS-730-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12632187
View Documents
Epredia
MS-730-P1
500 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12612187
View Documents
Epredia
MS-730-P
1 mL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
750.00 €
616.92 € / 1mL
Save 133.08 €
17% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12642187
View Documents
Epredia
MS-730-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12652187
View Documents
Epredia
MS-730-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12682187
View Documents
Epredia
MS-730-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: e2-4001 + 3B5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Cytoplasmic

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human HER-2 protein (3B5)

Molecular Weight: 185kDa

Positive Control: LS174T cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (For cytoplasmic)

Specifications

Specifications

HER-2/c-erbB-2/neu Ab-17
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ASR, IVD
Human, Rat
IgG1
e2-4001 + 3B5
Monoclonal
Mouse
EGFR Family
Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5)
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.