missing translation for 'onlineSavingsMsg'
Learn More

HERC3 Antibody [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30516539 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30516539 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30516539 Supplier Novus Biologicals Supplier No. NBP338562MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HERC3 Polyclonal antibody specifically detects HERC3 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2, HECT domain and RCC1-like domain-containing protein 3, hect domain and RLD 3, KIAA0032EC 6.3.2.-, probable E3 ubiquitin-protein ligase HERC3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human HERC3 (NP_055421.1).,, Sequence:, MLCWGYWSLGQPGISTNLQGIVAEPQVCGFISDRSVKEVACGGNHSVFLLEDGEVYTCGLNTKGQLGHEREGNKPEQIGALADQHIIHVACGESHSLALSDRGQLFSWGAGSDGQLGLMTTEDSVAVPRLIQKLNQQTILQVSCGNWHCLALAADGQFFTWGKNSHGQLGLGKEFPSQASPQRVRSLEGIPLAQVAAGGA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8916
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.