missing translation for 'onlineSavingsMsg'
Learn More

Hexabromobenzene, 97%

Product Code. 11480897
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480897 100 g 100g
11470897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480897 Supplier Thermo Scientific Alfa Aesar Supplier No. B23295.22
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 87-82-1 | C6Br6 | 551.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-82-1
Molecular Formula C6Br6
Molecular Weight (g/mol) 551.49
MDL Number MFCD00000058
InChI Key CAYGQBVSOZLICD-UHFFFAOYSA-N
Synonym hexabromobenzene, benzene, hexabromo, perbromobenzene, unii-t01859xwir, ccris 5917, benzene, 1,2,3,4,5,6-hexabromo, hxbbz, hexabioomobenzene, acmc-20aj0o
PubChem CID 6905
IUPAC Name 1,2,3,4,5,6-hexabromobenzene
SMILES C1(=C(C(=C(C(=C1Br)Br)Br)Br)Br)Br

Specifications

Melting Point >300°C
Odor Odorless
Quantity 100 g
Beilstein 1912586
Formula Weight 551.52
Percent Purity 97%
Chemical Name or Material Hexabromobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.