missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanedioic Acid Mono-(2-carboxyphenyl) Ester, Mikromol
Description
Hexanedioic Acid Mono-(2-carboxyphenyl) Ester, Mikromol
Specifications
Specifications
| Chemical Name or Material | Hexanedioic Acid mono-(2-Carboxyphenyl) Ester |
| CAS | 863985-44-8 |
| Analyte or Component Names | Hexanedioic Acid mono-(2-Carboxyphenyl) Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Hexanedioic acid 1-(2-carboxyphenyl) ester, Hexanedioic acid, mono(2-carboxyphenyl) ester (9CI) |
| InChI Formula | InChI=1S/C13H14O6/c14-11(15)7-3-4-8-12(16)19-10-6-2-1-5-9(10)13(17)18/h1-2,5-6H,3-4,7-8H2,(H,14,15)(H,17,18) |
| Solution Type | Neat |
| SMILES | OC(=O)CCCCC(=O)Oc1ccccc1C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?