missing translation for 'onlineSavingsMsg'
Learn More

hHR23b Antibody (SAIC-28B-8) - Azide and BSA Free, Novus Biologicals™

Product Code. 18653115 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.02mg
0.2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18653115 0.2 mg 0.2mg
18642454 0.025 mg 0.02mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18653115 Supplier Novus Biologicals Supplier No. NBP3200970.2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

hHR23b Monoclonal antibody specifically detects hHR23b in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen hHR23b
Applications Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry
Classification Monoclonal
Clone SAIC-28B-8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias HHR23B, HR23BUV excision repair protein RAD23 homolog B, p58, RAD23 (S. cerevisiae) homolog B, RAD23 homolog B (S. cerevisiae), RAD23, yeast homolog of, B, XP-C repair complementing complex 58 kDa, XP-C repair complementing protein, XP-C repair-complementing complex 58 kDa protein
Host Species Rabbit
Immunogen Peptide 'IDIDPEETVK', derived from the UV excision repair protein RAD23 homolog B, conjugated to KLH.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5887
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.