missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
hHR23b Monoclonal antibody specifically detects hHR23b in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
Specifications
Specifications
| Antigen | hHR23b |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry |
| Classification | Monoclonal |
| Clone | SAIC-28B-8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | HHR23B, HR23BUV excision repair protein RAD23 homolog B, p58, RAD23 (S. cerevisiae) homolog B, RAD23 homolog B (S. cerevisiae), RAD23, yeast homolog of, B, XP-C repair complementing complex 58 kDa, XP-C repair complementing protein, XP-C repair-complementing complex 58 kDa protein |
| Host Species | Rabbit |
| Immunogen | Peptide 'IDIDPEETVK', derived from the UV excision repair protein RAD23 homolog B, conjugated to KLH. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?