missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Human, Mouse, Rat, Fish, Hamster, Primate, Rabbit, Reptile, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In vitro assay, Gel Super Shift Assays, SDS-Page, Knockout Validated, Knockdown Validated, Dual RNAscope ISH-IHC.
Specifications
Specifications
| Classification | Polyclonal |
| Gene Alias | anti hif 2 alpha, anti-HIF-2 alpha, Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, |
| Gene Symbols | EPAS1 |
| Molecular Weight of Antigen | 96.5 kDa |
| Purification Method | Affinity Purified |
| Target Species | Rat, Hamster |
| Content And Storage | Store at -20 degreesC. |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?