missing translation for 'onlineSavingsMsg'
Learn More

HIF-2 alpha/EPAS1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18728423 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18728423 0.025 mL 25µL
18768873 - 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18728423 Supplier Novus Biologicals Supplier No. NB100122SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 663 publications

HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Human, Mouse, Rat, Fish, Hamster, Primate, Rabbit, Reptile, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In vitro assay, Gel Super Shift Assays, SDS-Page, Knockout Validated, Knockdown Validated, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-2 alpha/EPAS1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, Simple Western 1:50, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 5 ug/ 1 mg lysate, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28115701), In vitro assay reported in scientific literature (PMID 24998849), Gel Super Shift Assays reported in scientific literature (PMID 15184875), SDS-Page, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 26861754), KnockDown Validated reported in scientific literature (PMID 31061092), Dual RNAscope ISH-IHC
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q99814
Gene Alias anti hif 2 alpha, anti-HIF-2 alpha, Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor,
Gene Symbols EPAS1
Host Species Rabbit
Immunogen This HIF-2 alpha/EPAS1 Antibody was developed against a peptide derived from the C-terminus of mouse/human HIF-2 alpha protein.
Molecular Weight of Antigen 96.5 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2034
Test Specificity This HIF-2 alpha/EPAS1 Antibody is specific for HIF-2 alpha/EPAS, and does not cross-react with HIF-1 alpha.
Target Species Human, Mouse, Rat, Fish, Hamster, Primate, Rabbit, Reptile, Sheep
Content And Storage Store at -20 degreesC.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.