missing translation for 'onlineSavingsMsg'
Learn More

HIST1H2BC Antibody (CL5101) [Janelia Fluor« 549], Novus Biologicals Biologicals™

Product Code. 30530415 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30530415 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30530415 Supplier Novus Biologicals Supplier No. NBP344254JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HIST1H2BC Monoclonal antibody specifically detects HIST1H2BC in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIST1H2BC
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL5101
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias H2B histone family, member K, H2B/a, H2B/g, H2B/kH2B/h, H2BFA, H2BFG, H2BFH, H2BFKH2B/l, H2BFL, HIST1H2BC, HIST1H2BE, HIST1H2BF, HIST1H2BG, histone 1, H2bi, histone cluster 1, H2bi, histone H2B type 1-C/E/F/G/I, Histone H2B.1 A, Histone H2B.a, Histone H2B.g, Histone H2B.h, Histone H2B.k, Histone H2B.l
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P62807, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8347
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.