missing translation for 'onlineSavingsMsg'
Learn More

Histamine N-Methyltransferase/HNMT Antibody (034), PerCP, R&D Systems™

Product Code. 18613312 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18613312 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613312 Supplier Novus Biologicals Supplier No. NBP290171PCP
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Histamine N-Methyltransferase/HNMT Monoclonal specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for ELISA/Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histamine N-Methyltransferase/HNMT
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunoprecipitation
Gene Alias EC 2.1.1.8, histamine N-methyltransferase, HMT, HNMT-S1, HNMT-S2
Host Species Rabbit
Gene ID (Entrez) 3176
Content And Storage Store at 4C in the dark.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.