missing translation for 'onlineSavingsMsg'
Learn More

Histidine (L-Histidine), Mikromol™

Product Code. 17959814
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959814 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17959814 Supplier Mikromol Supplier No. MM1526.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Histidine (L-Histidine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Histidine
CAS 71-00-1
Analyte or Component Names L-Histidine
Shipping Condition Room Temperature
Molecular Formula C6 H9 N3 O2
Certifications/Compliance ISO 17034
Synonym Histidine, L-Histidine (ACI), Histidine, L- (8CI), (2S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid, (2S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid, (2S)-2-Azaniumyl-3-(1H-imidazol-5-yl)propanoate, (S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid, (S)-4-(2-Amino-2-carboxyethyl)imidazole, (S)-Histidine, (S)-α-Amino-1H-imidazole-4-propanoic acid, 103: PN: WO2021061872 SEQID: 103 claimed protein, 124: PN: WO2020191245 SEQID: 252 claimed protein, 177: PN: WO2021226558 SEQID: 252 claimed protein, 1H-Imidazole-4-alanine, (S)-, 1H-Imidazole-4-propanoic acid, α-amino-, (S)-, 8: PN: EP2878667 TABLE: 5 claimed sequence, L-(-)-Histidine, L-Alanine, 3-(1H-imidazol-4-yl)-, Glyoxaline-5-alanine, NSC 137773, (S)-2-Amino-3-(imidazol-4-yl)propanoic acid
InChI Formula InChI=1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1
SMILES N[C@@H](Cc1c[nH]cn1)C(=O)O
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid
Molecular Weight (g/mol) 155.15
Quantity 250 mg
Formula Weight 155.0695 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.