missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 4/HDAC4, p Ser632 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54636-0.025mg
This item is not returnable.
View return policy
Description
Histone Deacetylase 4/HDAC4 Polyclonal specifically detects Histone Deacetylase 4/HDAC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence.
Specifications
| Histone Deacetylase 4/HDAC4 | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% sodium azide | |
| HDAC4 | |
| HDAC4 (Ab-632) antibody was raised against a peptide sequence around aa. 630∼634 (A-Q-S-S-P) derived from Human HDAC4. | |
| 0.025 mg | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 | |
| HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9759 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction