missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Deacetylase 4/HDAC4, p Ser632 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
206.00 € - 781.00 €
Specifications
| Antigen | Histone Deacetylase 4/HDAC4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658496
|
Novus Biologicals
NBP2-54636-0.025mg |
0.025 mg |
206.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625526
|
Novus Biologicals
NBP2-54636 |
0.1 mg |
781.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone Deacetylase 4/HDAC4 Polyclonal specifically detects Histone Deacetylase 4/HDAC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence.Specifications
| Histone Deacetylase 4/HDAC4 | |
| Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 | |
| Polyclonal | |
| Rabbit | |
| HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
| HDAC4 | |
| IgG | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Human, Mouse, Rat | |
| 9759 | |
| HDAC4 (Ab-632) antibody was raised against a peptide sequence around aa. 630∼634 (A-Q-S-S-P) derived from Human HDAC4. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title