missing translation for 'onlineSavingsMsg'
Learn More

Histone H3, Dimethyl Lys4 Antibody (RM135), R&D Systems™

Product Code. 18621397 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18621397 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18621397 Supplier Novus Biologicals Supplier No. NBP261540
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP), Multiplex Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3
Applications Western Blot, ELISA, Immunofluorescence, ChIP Assay, Multiplex
Classification Monoclonal
Clone RM135
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.02 μg/mL - 0.2 μg/mL, ELISA 0.2 μg/mL - 1 μg/mL, Immunocytochemistry/ Immunofluorescence 1 μg/mL - 2 μg/mL, Chromatin Immunoprecipitation (ChIP) 1ug-5 μg, Multiplex Immunoassay 0.1 μg/mL - 0.5 μg/mL
Formulation 50% Glycerol/PBS, 1% BSA
Gene Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K4Me2, HIST2H3C, histone H3.3, MGC87782, MGC87783
Host Species Rabbit
Immunogen This Histone H3 [Dimethyl Lys4] antibody (RM135) was raised against a dimethyl-peptide corresponding to Histone H3 [Dimethyl Lys4]
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary or Secondary Primary
Gene ID (Entrez) 126961
Target Species Human, Vertebrates
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.