Learn More
HIV-1 tat Antibody - Azide and BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NB120-14029-0.025mg
 View more versions of this product
Product Code. 18575240
  • 336.00 € / 25µg
Description
HIV-1 tat Polyclonal specifically detects HIV-1 tat in Non-species specific samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HIV-1 tat
Polyclonal
Unconjugated
PBS with No Preservative
Chicken
Affinity Purified
RUO
Primary
Human
Western Blot
1 mg/mL
Western Blot 1:500-1:2000
Tat
Full length HIV-1 tat protein (Human).
0.025 mg
Virology Bacteria and Parasites
This may cross-react to various subtypes of HIV-1 tat protein
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.