missing translation for 'onlineSavingsMsg'
Learn More

HIV-1 tat Antibody - Azide and BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody

336.00 €

Specifications

Antigen HIV-1 tat
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18575240
View Documents
Novus Biologicals
NB120-14029-0.025mg
0.025 mg
336.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HIV-1 tat Polyclonal specifically detects HIV-1 tat in Non-species specific samples. It is validated for Western Blot.
Specifications

Specifications

HIV-1 tat
Western Blot 1:500-1:2000
Polyclonal
Chicken
Virology Bacteria and Parasites
PBS with No Preservative
Full length HIV-1 tat protein (Human).
Affinity Purified
This may cross-react to various subtypes of HIV-1 tat protein
1 mg/mL
Western Blot
Unconjugated
RUO
Human
Tat
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.