missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-A3 Monoclonal Antibody (GAP.A3), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ H071T03Y01-A
This item is not returnable.
View return policy
Description
HLA-A3 is a subtype of the HLA class I heavy chain paralogues, existing in two forms: HLA-A3.1 and HLA-A3.2, which differ at amino acid positions 152 and 156. HLA class I molecules are heterodimers consisting of a heavy chain and a light chain (beta-2 microglobulin), with the heavy chain anchored in the membrane. These molecules are highly polymorphic cell surface glycoproteins expressed on nearly all nucleated cells. HLA class I molecules play a central role in the immune system by presenting intracellular peptides derived from the endoplasmic reticulum lumen on the cell surface. Non-native peptides are recognized by circulating cytotoxic T lymphocytes, facilitating immune surveillance. The heavy chain of HLA-A is approximately 45 kDa and its gene contains 8 exons. Exons 2 and 3 encode the alpha1 and alpha2 domains, which bind the peptide, while exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exons 2 and 3 determine the peptide binding specificity of each class I molecule, and typing for these polymorphisms is routinely performed for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described, and HLA and MHC antibodies are crucial in immunopeptidomics, aiding in the identification and characterization of neoantigens through techniques such as high-performance liquid chromatography coupled to tandem mass spectrometry.
Specifications
| HLA-A3 | |
| Monoclonal | |
| 0.6 μg/Test | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P04439 | |
| HLA-A | |
| Affinity chromatography | |
| RUO | |
| 3105 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG2a κ |
| Flow Cytometry | |
| GAP.A3 | |
| NovaFluor Yellow 570 | |
| HLA-A | |
| A29 protein; antigen presenting molecule; Aw-33; Aw-74; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leukocyte antigen class I-A; major histocompatibility complex, class I, A; MCH class I antigen; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; truncated MHC class I antigen | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction