missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-A3 Monoclonal Antibody (GAP.A3), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
185.00 € - 392.00 €
Specifications
| Antigen | HLA-A3 |
|---|---|
| Clone | GAP.A3 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055148
|
Invitrogen™
H071T03Y01-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055765
|
Invitrogen™
H071T02Y01-A |
25 Tests |
185.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-A3 Monoclonal antibody specifically detects HLA-A3 in Human samples. It is validated for Flow CytometrySpecifications
| HLA-A3 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P04439 | |
| 3105 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| GAP.A3 | |
| Flow Cytometry | |
| NovaFluor Yellow 570 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A29 protein; antigen presenting molecule; Aw-33; Aw-74; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leukocyte antigen class I-A; major histocompatibility complex, class I, A; MCH class I antigen; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; truncated MHC class I antigen | |
| IgG2a κ | |
| Affinity chromatography | |
| HLA-A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title