missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HLA Class I Monoclonal antibody specifically detects HLA Class I in Human, Bovine, Primate samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
Specifications
Specifications
| Antigen | HLA Class I |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation, CyTOF |
| Classification | Monoclonal |
| Clone | MEM-123 |
| Conjugate | Biotin |
| Dilution | Flow Cytometry, ELISA, Immunoprecipitation, CyTOF-ready |
| Formulation | PBS |
| Gene Alias | A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 |
| Host Species | Mouse |
| Immunogen | COS-7 African green monkey kidney cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?