missing translation for 'onlineSavingsMsg'
Learn More

HLA DRA Antibody (169-1B5.2), Biotin, Novus Biologicals™

Product Code. p-200057461 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
100 μg
Unit Size:
0.1mL
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18627055 100 μg 100µL
18656655 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18627055 Supplier Novus Biologicals Supplier No. NBP247722B100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA DRA Monoclonal antibody specifically detects HLA DRA in Human, Bovine, Feline samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA DRA
Applications Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 169-1B5.2
Conjugate Biotin
Formulation PBS
Gene Alias FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
Host Species Mouse
Immunogen Human peripheral blood leukocytes (Uniprot: P01903)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3122
Target Species Human, Bovine, Feline
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.