missing translation for 'onlineSavingsMsg'
Learn More

HOMER1 Antibody, Novus Biologicals™

Product Code. 18205253 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18205253 0.025 mL 25µL
18767163 - 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18205253 Supplier Novus Biologicals Supplier No. NBP144999SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

HOMER1 Polyclonal specifically detects HOMER1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOMER1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. Q86YM7, Q9Z2Y3
Gene Alias HOMER, homer homolog 1 (Drosophila), homer-1, HOMER1B, HOMER-1B, HOMER1C, SYN47neuronal immediate early gene, 1
Gene Symbols HOMER1
Host Species Rabbit
Immunogen N-terminal partial recombinant human HOMER protein expressed in E. coli. [Swiss-Prot# Q86YM7]
Molecular Weight of Antigen 40 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 9456
Test Specificity Reacts with human and mouse. Immunogen sequence has 99% identity to rat, 98% to bovine, 96% to Xenopus and 74% to zebrafish.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.