missing translation for 'onlineSavingsMsg'
Learn More

HOXD4 Antibody (OTI1E9), DyLight 755, Novus Biologicals™

Product Code. 18112732 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18112732 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18112732 Supplier Novus Biologicals Supplier No. NBP270919IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HOXD4 Monoclonal specifically detects HOXD4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E9
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HHO.C13, homeo box D4, homeobox D4, Homeobox protein HHO.C13, Homeobox protein Hox-4B, Homeobox protein Hox-5.1, homeobox protein Hox-D4, HOX4, Hox-4.2, Hox-4.2, mouse, homolog of homeo box X, HOX4BHOX-5.1
Gene Symbols HOXD4
Host Species Mouse
Immunogen Full length recombinant protein of human HOXD4 (NP_055436) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3233.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.